missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chondrolectin Antibody (396305) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2576JF669
This item is not returnable.
View return policy
Description
Chondrolectin Monoclonal antibody specifically detects Chondrolectin in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF
Specifications
| Chondrolectin | |
| Monoclonal | |
| Janelia Fluor 669 | |
| C21orf68, chondrolectin, chromosome 21 open reading frame 68, FLJ12627, MT75, PRED12, Transmembrane protein MT75 | |
| Mouse myeloma cell line NS0-derived recombinant human Chondrolectin, Arg22-Asn216, Accession # Q9H9P2 | |
| 0.1 mL | |
| Developmental Biology | |
| 140578 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF | |
| 396305 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction