missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHORDC1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78304MFV610
This item is not returnable.
View return policy
Description
CHORDC1 Polyclonal antibody specifically detects CHORDC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CHORDC1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| CHORD domain-containing protein 1, CHORD-containing protein 1, CHP1CHP-1, cysteine and histidine-rich domain (CHORD) containing 1, cysteine and histidine-rich domain (CHORD)-containing 1, cysteine and histidine-rich domain (CHORD)-containing, zinc-binding protein 1, cysteine and histidine-rich domain-containing protein 1, FLJ37289, Protein morgana | |
| A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1] | |
| 0.1 mL | |
| Cancer, Epigenetics | |
| 26973 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction