Learn More
Chordin Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98688MFV610
This item is not returnable.
View return policy
Description
Chordin Polyclonal antibody specifically detects Chordin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Rabbit | |
| 8646 |