missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin alpha/beta (hCG) Antibody (HCGab/52), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11526H
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin alpha/beta (hCG) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha/beta (hCG) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Chorionic Gonadotropin alpha/beta (hCG) | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HCGab/52 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CGB;CGB3;CGB5;CGB7;CGB8;Choriogonadotropin subunit beta 3;Choriogonadotropin subunit beta 7;hCGB;LHB | |
| Purified Chorionic Gonadotropin alpha/beta (hCG) protein | |
| 0.1 mL | |
| Cancer | |
| 1082 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction