missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Antibody (LHa/756), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54500C
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Chorionic Gonadotropin alpha Chain (hCG alpha) | |
| Monoclonal | |
| DyLight 650 | |
| CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
| Recombinant full-length Chorionic Gonadotropin alpha Chain (hCG alpha) protein (Uniprot: P01215) | |
| 100 μg | |
| Cancer | |
| 1081 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LHa/756 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction