missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Rabbit anti-Human, Clone: HCGa/2728R, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 280.00 - € 528.00
Specifications
| Antigen | Chorionic Gonadotropin alpha Chain (hCG alpha) |
|---|---|
| Clone | HCGa/2728R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18339386
|
Novus Biologicals
NBP3-13844-20UG |
20 μg |
€ 280.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18389213
|
Bio-Techne
NBP3-13844-100UG |
100 μg |
€ 528.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Monoclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Chorionic Gonadotropin alpha Chain (hCG alpha) | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
| Recombinant fragment (around aa1-100) of human Chorionic Gonadotropin alpha Chain (hCG alpha) protein (exact sequence is proprietary) (Uniprot: P01215) | |
| Primary | |
| Store at 4°C. |
| HCGa/2728R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P01215 | |
| 1081 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title