missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63167AF594
This item is not returnable.
View return policy
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal antibody specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA
Specifications
| Chorionic Gonadotropin beta Chain (hCG beta) | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| INN-hCG-53 | |
| Western Blot, ELISA | |
| CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB | |
| Made to Human hCG holo C2 | |
| 0.25 mL | |
| Cancer | |
| 1082 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction