Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (INN-hCG-53), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-63167IR
Product Code. 18081614
  • ONLINE EXCLUSIVE PRICE
    € 593.00
    € 560.70 / 0.25mL
    Save € 32.30
    5% Off
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hCG holo C2
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CGB3
Made to Human hCG holo C2
0.25 mL
Cancer
1082
Human
Purified
Western Blot, ELISA
INN-hCG-53
Western Blot, ELISA
choriogonadotropin subunit beta, CGB3, CGB5, CGB7, CGB8, chorionic gonadotropin, beta polypeptide, hCGB
Mouse
Protein A or G purified
RUO
Primary
NB100-63167 recognizes epitope c2 on hCG holo C2 . It does not recognize free hCG subunits.
Store at 4C in the dark.