missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHP1 Monoclonal Antibody (OTI2E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810158
This item is not returnable.
View return policy
Description
This gene encodes a phosphoprotein that binds to the Na+/H+ exchanger NHE1. This protein serves as an essential cofactor which supports the physiological activity of NHE family members and may play a role in the mitogenic regulation of NHE1. The protein shares similarity with calcineurin B and calmodulin and it is also known to be an endogenous inhibitor of calcineurin activity.Specifications
| CHP1 | |
| Monoclonal | |
| Unconjugated | |
| CHP1 | |
| CHP, SLC9A1BP, Sid470p, p22, p24 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11261 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2E2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99653 | |
| CHP1 | |
| Full length human recombit protein of human CHP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction