missing translation for 'onlineSavingsMsg'
Learn More

CHRFAM7A Antibody [PerCP], Novus Biologicals Biologicals™

Product Code. 30513962 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30513962 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30513962 Supplier Novus Biologicals Supplier No. NBP338474PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CHRFAM7A Polyclonal antibody specifically detects CHRFAM7A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRFAM7A
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Gene Alias CHRNA7, CHRNA7 (cholinergic receptor, nicotinic, alpha 7, exons 5-10) and FAM7A (familywith sequence similarity 7A, exons A-E) fusion, CHRNA7 (cholinergic receptor, nicotinic, alpha polypeptide 7, exons 5-10) andFAM7A (family with sequence similarity 7A, exons A-E) fusion, CHRNA7-DR1alpha 7 neuronal nicotinic acetylcholine receptor-FAM7A hybrid, CHRNA7-FAM7A fusion protein, D-10alpha-7 nicotinic cholinergic receptor subunit, MGC120482, MGC120483
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human CHRFAM7A (NP_647536.1).,, Sequence:, MQKYCIYQHFQFQLLIQHLWIAANCDIADERFDATFHTNVLVNSSGHCQYLPPGIFKSSCYIDVRWFPFDVQHCKLKFGSWSYGGWSLDLQMQEADISGYIPNGEWDLVGIPGKRSERFYECCKEPYPDVTFTVTMRRRTLYYGLNLLIP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89832
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.