Learn More
CHRM3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21978-1-AP-150UL
 View more versions of this product
Product Code. 18038095
  • € 410.00 / 150µL
Description
The CHRM3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CHRM3. This antibody recognizes human, mouse, rat antigen. The CHRM3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CHRM3
400 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
CHRM3, HM3
150 μL
1131
Store at -20°C. Stable for one year after shipment.