Learn More
Invitrogen™ CHRM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550490
Product Code. 15694415
  • € 524.00 / 100µL
Description
The antibody detects endogenous levels of total CHRM4 protein.
Muscarinic acetylcholine receptor M4 is an Acetylcholine Receptor. Unlike muscarinic receptor M2, the M4 receptor is not involved in smooth muscle contraction. The clinical relevance of this receptor is unknown. However, mouse studies have linked it to adenylyl cyclase inhibition. Muscarinic acetylcholine receptor M4 has been reported to be expressed in many regions of the rat brain, particularly the striatum and hippocampus. ESTs have been isolated from human breast and eye libraries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CHRM4
Polyclonal
Unconjugated
CHRM4
acetylcholine receptor; acetylcholine receptor, muscarinic 4; AChR M4; cholinergic receptor; cholinergic receptor muscarinic 4; cholinergic receptor, muscarin 4; cholinergic receptor, muscarinic 4; CHRM4; Chrm-4; HM4; M4; M4R; Mm4 mAChR; muscarinic acetylcholine receptor 4; muscarinic acetylcholine receptor M4
Rabbit
Antigen affinity chromatography
RUO
1132, 12672, 25111
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P08173, P08485, P32211
CHRM4
Synthetic peptide corresponding to a region derived from internal residues of human Cholinergic receptor muscarinic 4.
100 μL
Primary
Human, Mouse, Rat