missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNA2 Mouse anti-Human, Rat, Clone:7G12H9, Abnova™
Mouse monoclonal antibody raised against recombinant human CHRNA2.
Brand: Abnova MAB17552.100ug
This item is not returnable.
View return policy
Specifications
| CHRNA2 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry Flow Cytometry Immunohistochemistry The optimal working dilution should be determined by the end user. |
|
| CHRNA2 | |
| Recombinant protein corresponding to amino acid 27-264 of human CHRNA2 from E. coli. | |
| 1135 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
| ELISA, Western Blot | |
| 7G12H9 | |
| cholinergic receptor, nicotinic, alpha 2 (neuronal) | |
| In PBS (0.05% sodium azide) | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction