missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNA5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540994
This item is not returnable.
View return policy
Description
Peptide sequence: PDIVLFDNAD GRFEGTSTKT VIRYNGTVTW TPPANYKSSC TIDVTFFPFD Sequence homology: Cow: 85%; Dog: 79%; Horse: 85%; Human: 100%; Mouse: 92%; Pig: 85%; Rabbit: 92%; Rat: 92%.
Nicotinic acetylcholine receptors (nAChRs), such as CHRNA5, are members of a superfamily of ligand-gated ion channels that mediate fast signal transmission at synapses. The nAChRs are thought to be (hetero)pentamers composed of homologous subunits.
Specifications
| CHRNA5 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA5 | |
| acetylcholine receptor; acetylcholine receptor alpha 5; acetylcholine receptor, nicotinic, alpha 5 (neuronal); Acra5; Acra-5; cholinergic receptor; cholinergic receptor nicotinic alpha 5 subunit; Cholinergic receptor, neuronal nicotinic, alpha polypeptide-5; cholinergic receptor, nicotinic alpha 5; cholinergic receptor, nicotinic, alpha 5; cholinergic receptor, nicotinic, alpha 5 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 5; CHRNA5; hypothetical protein LOC550584; LNCR2; nAChR alpha 5; NACHRA5; Neuronal acetylcholine receptor subunit alpha-5; neuronal nicotinic acetylcholine receptor alpha5 subunit; neuronal nicotinic acetylcholine receptor, alpha5 subunit; nicotinic acetylcholine receptor alpha5 subunit; zgc:110642 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P30532 | |
| CHRNA5 | |
| Synthetic peptide directed towards the middle region of human CHRNA5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction