missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNA6 Monoclonal Antibody (5B6G8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531689
This item is not returnable.
View return policy
Description
MA5-31689 has been tested in indirect ELISA.
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
Specifications
| CHRNA6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q15825 | |
| CHRNA6 | |
| Purified recombinant fragment of human CHRNA6 (AA: 26-239) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5B6G8 | |
| Unconjugated | |
| CHRNA6 | |
| acetylcholine receptor; acetylcholine receptor alpha 6 neural; acetylcholine receptor, nicotinic, alpha 6 (neuronal); Acra6; alpha6 nAChR; CHNRA6; cholinergic receptor; cholinergic receptor nicotinic alpha 6 subunit; cholinergic receptor, nicotinic alpha 6; cholinergic receptor, nicotinic, alpha 6 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 6; CHRNA6; Neuronal acetylcholine receptor subunit alpha-6; Nica6; nicotinic acetylcholine receptor subunit alpha 6 | |
| Mouse | |
| Protein G | |
| RUO | |
| 8973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction