missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115651
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Nicotinic Acetylcholine Receptor alpha 7.
Chromosome 15 encodes over 700 genes and comprises nearly 3% of the human genome. CHRFAM7A, a fusion protein comprised of FAM7A and AChR alpha7, is encoded by the CHRFAM7A gene, which is located on a region of human chromosome 15 that is associated with several neuropsychiatric disorders, such as schizophrenia and bipolar affective disorder. CHRFAM7A, also known as D-10 or CHRNA7, is a 412 amino acid multi-pass membrane protein that belongs to the nicotinic acetylcholine receptor (nAChR) subfamily of ligand-gated ion channel proteins, a protein superfamily that mediates signal transmission at synapses. CHRFAM7A is expressed in the hippocampus. Alternative splicing of the CHRFAM7A gene produces two splice variants.
Specifications
| CHRNA7 | |
| Polyclonal | |
| Unconjugated | |
| CHRNA7 | |
| a7 nicotinic acetylcholine receptor; acetylcholine receptor; acetylcholine receptor alpha 7 neural; Acra7; alpha 7 neuronal nicotinic acetylcholine receptor; alpha7; alpha-7 nicotinic cholinergic receptor subunit; alpha7 nicotinic receptor; alpha7-nAChR; BTX; bungarotoxin alpha; C holinergic receptor nicotinic alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); C holinergic receptor, nicotinic, alpha polypeptide 7 (neuronal nicotinic acetycholine receptor alpha 7) (bungarotoxin alpha); cholinergic receptor; cholinergic receptor nicotinic alpha 7 subunit; cholinergic receptor, nicotinic alpha 7; cholinergic receptor, nicotinic, alpha 7 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 7; CHRFAM7A; Chrna7; CHRNA7-2; CHRNA7-DR1; D-10; nAchR; nAChR alpha7; NACHRA7; NARAD; neuronal acetylcholine receptor protein, alpha-7 chain; neuronal acetylcholine receptor subunit alpha-7; neuronal nicotinic acetycholine receptor alpha 7; nicotinic receptor alpha 7 subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1139, 11441, 25302 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36544, P49582, Q05941 | |
| CHRNA7 | |
| A synthesized peptide derived from human CHRNA7(Accession P36544), corresponding to amino acid residues H18-K68. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction