missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHRNB1 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518924
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
The muscle acetylcholine receptor is composed of five subunits: two alpha subunits and one beta, one gamma, and one delta subunit. This gene encodes the beta subunit of the acetylcholine receptor. The acetylcholine receptor changes conformation upon acetylcholine binding leading to the opening of an ion-conducting channel across the plasma membrane. Mutations in this gene are associated with slow-channel congenital myasthenic syndrome.
Specifications
| CHRNB1 | |
| Polyclonal | |
| Unconjugated | |
| Chrnb1 | |
| acetylcholine receptor; acetylcholine receptor beta; acetylcholine receptor subunit beta; acetylcholine receptor, nicotinic, beta 1 (muscle); AChR beta; Achr-2; ACHRB; Acrb; cholinergic receptor; cholinergic receptor nicotinic beta 1 subunit; cholinergic receptor nicotinic beta polypeptide 1; cholinergic receptor, nicotinic beta 1; cholinergic receptor, nicotinic, beta 1 (muscle); cholinergic receptor, nicotinic, beta polypeptide 1 (muscle); CHRNB; Chrnb 1; CHRNB1; CMS1D; CMS2A; CMS2C; nAChRB1; RNACRB1; SCCMS | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 1140 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P11230 | |
| Chrnb1 | |
| Synthetic peptide sequence (QEQEDHDALKED) corresponding to the internal amino acids of CHRNB1 (aa 448-460). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction