Learn More
CHRNB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17844-1-AP-150UL
 View more versions of this product
Product Code. 16827844
  • € 410.00 / 150µL
Description
Neuronal acetylcholine receptors are homo- or heteropentameric complexes composed of homologous alpha and beta subunits. They belong to a superfamily of ligand-gated ion channels which allow the flow of sodium and potassium across the plasma membrane in response to ligands such as acetylcholine and nicotine. This gene encodes one of several beta subunits. Mutations in this gene are associated with autosomal domit nocturnal frontal lobe epilepsy.
Specifications
Specifications
CHRNB2
Polyclonal
Unconjugated
CHRNB2
CHRNB2, EFNL3, nAChRB2
Rabbit
Antigen Affinity Chromatography
RUO
1141, 11444, 54239
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P12390, P17787, Q9ERK7
Chrnb2
CHRNB2 Fusion Protein Ag12208
150 μL
Primary
Human, Mouse, Rat