missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNB3 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97039AF594
This item is not returnable.
View return policy
Description
CHRNB3 Polyclonal specifically detects CHRNB3 in Human samples. It is validated for ELISA.
Specifications
| CHRNB3 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Alexa Fluor 594 | |
| acetylcholine receptor, neuronal nicotinic, beta-3 subunit, cholinergic receptor, nicotinic, beta 3, cholinergic receptor, nicotinic, beta polypeptide 3, neuronal acetylcholine receptor subunit beta-3 | |
| Produced in rabbits immunized with purified, recombinant CHRNB3 (NP_000740.1; Met1-Leu232). | |
| 0.1 mL | |
| Protein Phosphatase | |
| 1142 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction