missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHRNB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | CHRNB3 |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18602462
|
Novus Biologicals
NBP2-97039-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660865
|
Novus Biologicals
NBP2-97039-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHRNB3 Polyclonal antibody specifically detects CHRNB3 in Human samples. It is validated for ELISASpecifications
| CHRNB3 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Protein Phosphatase | |
| 0.2 um filtered solution in PBS | |
| 1142 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| acetylcholine receptor, neuronal nicotinic, beta-3 subunit, cholinergic receptor, nicotinic, beta 3, cholinergic receptor, nicotinic, beta polypeptide 3, neuronal acetylcholine receptor subunit beta-3 | |
| Produced in rabbits immunized with purified, recombinant Human CHRNB3 (Accession#: NP_000740.1; Met1-Leu232) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title