missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHRNB4 Polyclonal Antibody

Product Code. 16598481
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16598481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16598481 Supplier Invitrogen™ Supplier No. PA595643

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antigen Affinity Purified from Pooled Serum.

After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNB4
Applications Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation 10mM HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene CHRNB4
Gene Accession No. P12392, Q8R493
Gene Alias acetylcholine receptor; Acetylcholine receptor beta 4; acetylcholine receptor, nicotinic, beta 4 (neuronal); Acrb4; Acrb-4; cholinergic receptor; cholinergic receptor nicotinic beta 4 subunit; cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; Chrnb4; nAChRB4; N-alpha 2; neuronal acetylcholine receptor non-alpha-2 chain; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit; nicotinic acetylcholine receptor beta4 subunit
Gene Symbols CHRNB4
Host Species Rabbit
Immunogen Fusion protein from the cytoplasmic loop of the beta 4 subunit of rat nAChR.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108015, 25103
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.