Learn More
Macherey-Nagel™ Chromabond™ Easy Polypropylene SPE Column
Shop All Macherey-Nagel Products
missing translation for 'orderingAttributeHoverText'
Bed Weight:
30 mg
60 mg
100 mg
200 mg
Column Capacity (Metric):
1 mL
3 mL
6 mL
missing translation for 'unitSize'
Pack of 30
Specifications
Specifications
| Column Capacity (Metric) | 1 mL |
| Column Type | SPE |
| Bed Weight | 30 mg |
| For Use With (Application) | Solid Phase Extraction |
| Particle Size | 80 μm |
| Sorbent | Chromabond Easy |
| Type | Solid Phase Extraction Column |
| Pore Size | 50 μm |
| Surface Area (Metric) | 650 to 700 m2/g |