Learn More
Macherey-Nagel™ Chromabond Florisil Adsorbent
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Quantity | 100 g |
| For Use With (Application) | Organic Tin Compounds, Aliphatic Carboxylic Acids, PCBs, PAHs |
| Product Type | Chromabond Florisil Adsorbent |
| Particle Size | 150 to 250 μm |