Learn More
Macherey-Nagel™ Chromabond™ HR-P Polypropylene SPE Column
Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
100 mg
200 mg
500 mg
1 g
Column Capacity (Metric):
1 mL
3 mL
6 mL
Unit Size:
Each
Pack of 30
Specifications
Specifications
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| Bed Weight | 200 mg |
| Particle Size | 50 to 100 μm |
| Type | Solid Phase Extraction Column |
| Surface Area (Metric) | 1200 m2/g |