Learn More
Macherey Nagel X20 CHROMABOND Leather Columns Volume: 30ml .
Brand:  Macherey Nagel 730380.100.119600
X20 CHROMABOND Leather Columns Volume: 30ml .
Product Code. 10138544
  • ONLINE EXCLUSIVE PRICE
    € 1510.00
    € 1373.00 / Pack of 20
    Save € 137.00
    9% Off
Only null left
Estimated Shipment: 19-01-2026