Learn More
Macherey-Nagel™ Chromabond™ SA/SiOH Polypropylene SPE Column
Product Code. 12823941 Shop All Macherey-Nagel Products
Click to view available options
Column Capacity (Metric):
3 mL
Unit Size:
Pack of 50
Product Code. 12823941
Brand: Macherey-Nagel™ 730132
Only null left
Specifications
Base Material Silica
Column Capacity (Metric) 3 mL
Column Type SPE
For Use With (Application) Solid Phase Extraction Analysis of PCBs
Sorbent Chromabond SA/SiOH
Type Solid Phase Extraction Column