Learn More
Macherey-Nagel™ Chromabond™ SA/SiOH Polypropylene SPE Column
Shop All Macherey-Nagel Products
Click to view available options
Column Capacity (Metric):
3 mL
Unit Size:
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Column Capacity (Metric) | 3 mL |
| Column Type | SPE |
| For Use With (Application) | Solid Phase Extraction Analysis of PCBs |
| Sorbent | Chromabond SA/SiOH |
| Type | Solid Phase Extraction Column |