Learn More
Macherey-Nagel™ CHROMABOND SPE Vacuum Manifold
Product Code. 16099311 Shop All Macherey-Nagel Products
Change view
Click to view available options
No. of Ports:
12
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Ports Unit Size
16099311 12 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16099311 Supplier Macherey-Nagel™ Supplier No. 730150N
  • ONLINE EXCLUSIVE PRICE
    € 1045.00
    € 1034.25 / Pack of 1
    Save € 10.75
    1% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The complete CHROMABOND SPE vacuum manifold is the perfect choice for speeding up sample processing of up to 12 positions simultaneously.
  • Complete vacuum manifold system
Specifications
Description Vacuum Manifold, Complete System
For Use With (Application) Sample Preparation
Includes 1x Glass cabinet, 1x lid (complete, incl. 4x feet, Luer fittings, 1x lid gasket), 12x exchangeable needles (PP), 1x vacuum gauge, 1x control valve, 12x Luer valves (stopcocks), 12x caps, 1x variable rack (6 plates), 1x PP tank
No. of Ports 12