Learn More
Chromacol PVDF Syringe Filters
Thermo Scientific™ Chromacol PVDF Syringe Filters are offered in a seletion of diameter and pore sizes.
Brand: Thermo Scientific Chromacol 30-SF-45(PV)
216.91 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ Chromacol PVDF Syringe Filters are offered in a selection of diameters and pore sizes.
Specifications
PVDF | |
0.45 μm | |
Syringe Filter | |
30 mm |