missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Chroman 1
GREENER_CHOICE

Product Code. 18725466
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18725466

Brand: Tocris Bioscience™ 7163/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent and selective ROCK 2 inhibitor; improves cell survival after cryogeneisis

Chroman 1 is a highly potent and selective ROCK2 (Rho-kinase) inhibitor (IC50 values are 1 and 52 pM at ROCK2 and ROCK1, respectively). Chroman 1 exhibits >2000-fold selectivity for ROCK2 over a range of related kinases including MRCK, PKA, and AKT1 (IC50 values are 150, >20000, and >20000 nM, respectively). Chroman 1 promotes survival of pluripotent stem cells (PSCs) in culture when used in combination with Emricasan (Cat. No. 7310), Polyamines, and Trans-ISRIB (Cat. No. 5284), a combination known as CEPT. When used as part of the CEPT cocktail, Chroman 1 also improves survival of differentiated cells following cryopreservation with cardiomyocyte cell survival increasing by 36% and motor neuron survival improving by 63% compared with DMSO controls.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 S)-N-[2-[2-(Dimethylamino)ethoxy]-4-(1 H-pyrazol-4-yl)phenyl]-3,4-dihydro-6-methoxy-2 H-1-benzopyran-3-carboxamide dihydrochloride
Quantity 50 mg
Target Rho-kinase Inhibitors
Molecular Formula C24H28N4O40.2 HCl
Formula Weight 12373
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.