missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium AA Standard: Cr @ 1000 μg/mL in 5% HCl, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
Chromium AA Standard: Cr @ 1000 μg/mL in 5% HCl
Specifications
Specifications
| Chemical Name or Material | Chromium |
| CAS | 7440-47-3 |
| Methods | AA/GFAA |
| Analyte or Component Names | Chromium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Cr |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Cr |
| InChI Formula | InChI=1S/Cr |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction