missing translation for 'onlineSavingsMsg'
Learn More

Chromogranin A Antibody (CGA/413), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-47850DL594

Product Code. 18727261

  • ONLINE EXCLUSIVE PRICE
    € 573.00
    € 541.80 / 0.10mL
    Save € 31.20
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Chromogranin A Monoclonal antibody specifically detects Chromogranin A in Human, Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein Array
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chromogranin A
Monoclonal
DyLight 594
betagranin (N-terminal fragment of chromogranin A), CGA, chromogranin A (parathyroid secretory protein 1), chromogranin-A, parathyroid secretory protein 1, Pituitary secretory protein I, SP-I
Recombinant full-length human Chromogranin A protein (Uniprot: P10645)
0.1 mL
Apoptosis, Cancer, Neuronal Cell Markers, Tumor Biomarkers
1113
Store at 4°C in the dark.
IgG2b κ
Immunohistochemistry, Immunohistochemistry (Paraffin), Microarray, Protein Array
CGA/413
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Rat (Negative)
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.