missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A / CHGA Mouse, Clone: LK2H10 + PHE5 + CGA/414, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1113-MSM5-P1
This item is not returnable.
View return policy
Specifications
| Chromogranin A / CHGA | |
| Finely granular cytoplasmic | |
| LK2H10 + PHE5 + CGA/414 | |
| Unconjugated | |
| NP_001266 | |
| Mouse | |
| 68-75kDa | |
| Protein A/G Purified | |
| RUO | |
| 1113 | |
| IgG |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| CHGA | |
| Human pheochromocytoma (LK2H10 and PHE5) and recombinant human chromogranin A protein (CGA414) | |
| PC12 cells. Adrenal gland, bowel, thyroid, pancreas, or pheochromocytoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction