Learn More
Chromogranin B Antibody (CL11404) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15119PE
Product Code. 30302769
  • € 737.00 / 0.10mL
Description
Chromogranin B Monoclonal antibody specifically detects Chromogranin B in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chromogranin B
Monoclonal
PE
CgB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI
This antibody was developed using a recombinant protein derived from P05060, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cytokines and Growth Factors, Neuroscience, Signal Transduction
1114
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11404
PBS
Mouse
Protein A purified