missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin C Antibody (SGII 6B1/3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76658H
This item is not returnable.
View return policy
Description
Chromogranin C Monoclonal specifically detects Chromogranin C in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Chromogranin C | |
| Monoclonal | |
| HRP | |
| PBS with no preservative | |
| SCG2 | |
| Peptide from rat Chromogranin C (C terminus of p18: KEENSRENPF) | |
| Neuroscience | |
| 7857 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| SGII 6B1/3 | |
| Western Blot, Immunohistochemistry | |
| CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction