missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin C Antibody (SGII 6B1/3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76658MFV610
This item is not returnable.
View return policy
Description
Chromogranin C Monoclonal antibody specifically detects Chromogranin C in Rat samples. It is validated for Flow Cytometry
Specifications
| Chromogranin C | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry | |
| SGII 6B1/3 | |
| Flow Cytometry | |
| CHCG, Chromogranin-C, EM66, SCG2, secretogranin IICHGCchromogranin-C, secretoneurin, SgIIsecretogranin-2, SN | |
| Peptide from rat Chromogranin C (C terminus of p18: KEENSRENPF) | |
| 0.1 mL | |
| Neuroscience | |
| 7857 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction