Learn More
Proteintech ChromoTek Halo-Trap Agarose Beads Kit
Description
- Halo-Trap Agarose are affinity beads for IP of Halo-tag fusion proteins
- Comprises a Halo-tag Nanobody /VHH conjugated to agarose beads
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |