Learn More
Proteintech ChromoTek Halo-Trap Magnetic Agarose
Description
- Halo-Trap Magnetic Agarose is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the presence or absence of a covalently bound ligand
- The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible
Specifications
Specifications
| Binding Capacity | 12.5 μg of recombinant Halo-tag per 25 μL bead slurry |
| Content And Storage | Upon receipt store at 4°C. Do not freeze. Stable for 1 year. Shipped at ambient temperature. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Halo-tag (modified variant of the bacterial haloalkane dehalogenase enzyme from Rhodococcus rhodochrous) in the absence or presence of covalently bound chloralkane-based ligands |
| Label or Dye | Magnetic Agarose |
| Species | Alpaca |
| Sufficient For | 100 Reactions |