Learn More
Proteintech ChromoTek Myc-Trap™ Agarose
Description
- Consists of a Myc Nanobody/VHH coupled to agarose beads
Specifications
Specifications
| Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
| For Use With (Application) | IP, CoIP, ChIP, RIP |
| Target | Myc-tag sequence motif EQKLISEEDL at the N-terminus, C-terminus, or internal site of the fusion protein. Endogenous c-myc is NOT bound. |
| Label or Dye | Agarose |
| Species | Alpaca |
| Sufficient For | 20 Reactions |