Learn More
Proteintech ChromoTek Spot-Cap™ and Peptide
Product Code. 17393293
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17393293 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17393293 Supplier Proteintech Supplier No. ecaep
Slurry of Spot-Cap™ (eca-0.1) plus 1 mg Spot-peptide (ep-1).
  • Spot-Cap™ is a Nanobody-based affinity resin optimized for one-step protein purification of Spot-tagged proteins.