Learn More
Invitrogen™ CHST10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA592187
Description
Immunogen sequence: KDPDVYSAKQ EFLFLTTMPE VRKLPEEKHI PEELKPTGKE LPDSQLVQPL VYMERLELIR NVCRDDALKN LSHTPVSKFV LDRIFVCDKH KILFCQTPKV GNTQWKKVLI VLNGAFSSIE EIPENVVHDH EKNGLPRLSS FSDAEIQKRL KTYFKFFIVR DPFERLISAF KDKFVHNPRF EPWYRHEIAP GIIRKYRRNR TETRGIQFED FVRYLGDPNH R; Positive Samples: BxPC-3; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.[provided by RefSeq, Feb 2011].
Specifications
| CHST10 | |
| Polyclonal | |
| Unconjugated | |
| CHST10 | |
| AI507003; AU041319; carbohydrate sulfotransferase 10; CHST10; HNK-1 sulfotransferase; HNK1ST; HNK-1ST; huHNK-1ST; raHNK-1ST; ST; sul-T | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9486 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O43529 | |
| CHST10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-250 of human CHST10 (NP_0048451). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.