missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHST2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564950
This item is not returnable.
View return policy
Description
Immunogen sequence: HALGAMEVICN SMAKTLQTAL QPPDWLQGHY LVVRYEDLVG DPVKTLRRVY DFVGLLVSPE MEQFALNMTS GSGSSSKPFV VSARNATQAA NAWRTALTFQ QIKQVEEFCY QPMAVLGYER VNSPEEVKDL SKT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.
N-acetylglucosamine-6-O-sulfotransferases, such as CHST2, catalyze the transfer of sulfate from 3-prime-phosphoadenosine 5-prime-phosphosulfate.
Specifications
| CHST2 | |
| Polyclonal | |
| Unconjugated | |
| CHST2 | |
| AI428561; AW121776; C130041E03Rik; C6ST; carbohydrate (chondroitin 6/keratan) sulfotransferase 2; carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2; carbohydrate sulfotransferase 2; Chst2; Chts2; epididymis secretory protein Li 75; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2; GlcNAc6ST; glcNAc6ST-1; GN6ST; Gn6ST-1; GST2; GST-2; HEL-S-75; N-acetylglucosamine 6-O-sulfotransferase 1; N-acetylglucosamine-6-O-sulfotransferase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9435 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y4C5 | |
| CHST2 | |
| Recombinant Human CHST2. Recombinant protein control fragment (Product #RP-90506). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction