Learn More
CHT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21848-1-AP-20UL
 View more versions of this product
Product Code. 16891395
  • € 145.00 / 20µL
Description
Acetylcholine functions as a neurotransmitter in both the central and peripheral nervous systems of all vertebates, and is the principle neurotransmitter used at the neuromuscular junction. Presynaptic synthesis of acetylcholine requires a steady supply of choline, which is acquired by a plasma membrane high affinity choline transporter (CHT).
Specifications
Specifications
CHT1
Polyclonal
Unconjugated
SLC5A7
CHT, CHT1, hCHT, SLC5A7
Rabbit
Antigen Affinity Chromatography
RUO
60482, 63993, 85426
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.46 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BGY9, Q9GZV3, Q9JMD7
SLC5A7
CHT1 Fusion Protein Ag16136
20 μL
Primary
Human, Rat, Mouse