missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFAP3 Antibody (OTI1B3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72657F
This item is not returnable.
View return policy
Description
MFAP3 Monoclonal specifically detects MFAP3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MFAP3 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| MFAP3 | |
| Full length recombinant protein of human MFAP3 (NP_005918) produced in HEK293T cells. | |
| 0.1 mL | |
| 4238.0 | |
| Store at 4°C in the dark. |
| Immunocytochemistry, Immunofluorescence | |
| OTI1B3 | |
| Immunocytochemistry/Immunofluorescence | |
| DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction