missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHURC1 Monoclonal Antibody (2F9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524502
This item is not returnable.
View return policy
Description
Peptide Sequence: MCGDCVGKEY PNRGNTCLEN GSFLLNFTGC AVCSKRDFML ITNKSLKEED GEEIVTYDHL CKNCHHVIAR HEYTFSIMDE FQEYTMLCLL CGKAEDTISI LPDDPRQMTL LF.
Transcriptional activator that mediates FGF signaling during neural development. Plays a role in the regulation of cell movement. Does not bind DNA by itself.
Specifications
| CHURC1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8WUH1 | |
| CHURC1 | |
| CHURC1 (AAH20550, 1 a.a. ~ 112 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F9 | |
| Unconjugated | |
| CHURC1 | |
| 1700030P15Rik; C14orf52; CHCH; CHURC1; churchill domain containing 1; My015; Protein Churchill | |
| Mouse | |
| Protein A | |
| RUO | |
| 91612 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction