missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chymotrypsin C/CTRC Antibody (102), HRP , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89901H
This item is not returnable.
View return policy
Description
Chymotrypsin C/CTRC Monoclonal antibody specifically detects Chymotrypsin C/CTRC in Human samples. It is validated for ELISA
Specifications
| Chymotrypsin C/CTRC | |
| Monoclonal | |
| HRP | |
| Caldecrin, chymotrypsin C (caldecrin), chymotrypsin-C, CLCRserum calcium decreasing factor, EC 3.4.21, EC 3.4.21.2, ELA4, elastase 4, elastase IV | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Chymotrypsin C/CTRC (Q99895; Met1-Leu268). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 102 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 11330 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction