missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chymotrypsin-like protease Antibody (SR1373), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22235-100ul
This item is not returnable.
View return policy
Description
Chymotrypsin-like protease Monoclonal antibody specifically detects Chymotrypsin-like protease in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Chymotrypsin-like protease | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1373 | |
| Western Blot 1:500-1:2000 | |
| chymotrypsin-like, chymotrypsin-like protease CTRL-1, CTRL1, EC 3.4.21, EC 3.4.21.-, MGC70821 | |
| A synthesized peptide derived from human Chymotrypsin-like protease (Uniprot #: P40313) | |
| 100 μg | |
| Cell Biology | |
| 1506 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction