missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIAO1 Antibody (6C9), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42631DL594
This item is not returnable.
View return policy
Description
CIAO1 Monoclonal antibody specifically detects CIAO1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CIAO1 | |
| Monoclonal | |
| DyLight 594 | |
| CIA1, cytosolic iron-sulfur protein assembly 1, cytosolic iron-sulfur protein assembly 1 homolog (S. cerevisiae), probable cytosolic iron-sulfur protein assembly protein CIAO1, WD repeat domain 39, WD repeat-containing protein 39, WD40 protein Ciao1, WDR39cytosolic iron-sulfur protein assembly 1 homolog | |
| Recombinant human CIAO1 (1-339aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 9391 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 6C9 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction