missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP-1/HIAP-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00
Specifications
| Antigen | cIAP-1/HIAP-2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
cIAP-1/HIAP-2 Polyclonal specifically detects cIAP-1/HIAP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| cIAP-1/HIAP-2 | |
| Western Blot 1-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 10 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Apoptosis | |
| PBS with 0.02% Sodium Azide | |
| API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, cIAP1, C-IAP1, hIAP2, hIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2 | |
| BIRC2 | |
| IgG | |
| Affinity Purified | |
| 70 kDa |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q13490 | |
| 329 | |
| Antibody was raised against a synthetic peptide corresponding to 14 amino acids at the C-terminus of human c-IAP1. c-IAP antibody detects both c-IAP1 and c-IAP2. The immunogen is located within amino acids 540 - 590 of cIAP. Amino Acid Squence: SLEEQLRRLQEERT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.