missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cIAP-2/HIAP-1 Antibody (OTI1A9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70184AF532
This item is not returnable.
View return policy
Description
cIAP-2/HIAP-1 Monoclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| cIAP-2/HIAP-1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| BIRC3 | |
| Recombinant protein fragment corresponding to amino acids 255-529 of human cIAP-2/HIAP-1 (NP_001156) produced in E.coli. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI1A9 | |
| Western Blot, Flow Cytometry | |
| AIP1, API2, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49 | |
| Mouse | |
| 0.1 mL | |
| 330 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction