missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIAPIN1 Antibody (OTI5A6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72072FR
This item is not returnable.
View return policy
Description
CIAPIN1 Monoclonal specifically detects CIAPIN1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CIAPIN1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| CIAPIN1 | |
| Recombinant protein fragment corresponding to amino acids 97-312 of human CIAPIN1 (NP_064709) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5A6 | |
| Western Blot, Immunohistochemistry | |
| Anamorsin, cytokine induced apoptosis inhibitor 1, Cytokine-induced apoptosis inhibitor 1, DRE2, Fe-S cluster assembly protein DRE2 homolog, predicted protein of HQ0915,2810413N20Rik, PRO0915 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis | |
| 57019.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction